This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06780343_m1
See other MYLK4 GE Assays ›
Gene Symbol:
MYLK4
Gene Name:
myosin light chain kinase family member 4
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 2813610 - 2928723 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06780343_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYLK4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008274287.2 XP_008272509.1 2449 75
XM_008274288.2 XP_008272510.1 3013 75
XM_008274289.2 XP_008272511.1 2113 75
XM_008274290.2 XP_008272512.1 3253 75
XM_008274291.2 XP_008272513.1 2524 75
XM_008274292.2 XP_008272514.1 2623 75
XM_008274293.2 XP_008272515.2 1303 75
XM_017350515.1 XP_017206004.1 2053 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Oryctolagus cuniculus

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0