Assay Details
Gene Symbol: SERINC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714795.3 | XP_002714841.1 | 3 - 4 | 468 | 68 | |
XM_017345341.1 | XP_017200830.1 | 4 - 5 | 573 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
