This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06768129_m1
See other FGFR1OP2 GE Assays ›
Gene Symbol:
FGFR1OP2
Gene Name:
FGFR1 oncogene partner 2
Gene Aliases:
-
Chromosome Location:
Chr.8: 13198362 - 13240880 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06768129_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGFR1OP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002712698.3 XP_002712744.1 2 - 3 519 79
XM_008259494.2 XP_008257716.1 2 - 3 392 79
XM_008259495.2 XP_008257717.1 2 - 3 275 79
XM_008259496.2 XP_008257718.1 2 - 3 390 79
XM_008259497.2 XP_008257719.1 2 - 3 397 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0