Assay Details
Gene Symbol: SLC16A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274237.2 | XP_008272459.1 | 611 | 83 | ||
XM_008274238.2 | XP_008272460.1 | 757 | 83 | |||
XM_008274239.2 | XP_008272461.1 | 763 | 83 | |||
XM_008274240.2 | XP_008272462.1 | 768 | 83 | |||
XM_008274241.2 | XP_008272463.1 | 668 | 83 | |||
XM_008274242.2 | XP_008272464.1 | 717 | 83 | |||
XM_008274244.2 | XP_008272466.1 | 663 | 83 | |||
XM_008274245.2 | XP_008272467.1 | 768 | 83 | |||
XM_008274246.2 | XP_008272468.1 | 657 | 83 | |||
XM_008274247.2 | XP_008272469.1 | 629 | 83 | |||
XM_008274250.2 | XP_008272472.1 | 588 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
