This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06759725_m1
See other AARS1 GE Assays ›
Gene Symbol:
AARS1
Gene Name:
alanyl-tRNA synthetase 1
Gene Aliases:
AARS
Chromosome Location:
Chr.5: 29194388 - 29214152 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06759725_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008257608.2 XP_008255830.1 7 - 8 1011 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0