This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06744809_m1
See other TUT4 GE Assays ›
Gene Symbol:
TUT4
Gene Name:
terminal uridylyl transferase 4
Gene Aliases:
ZCCHC11
Chromosome Location:
Chr.13: 114146586 - 114288659 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06744809_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TUT4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008265237.2 XP_008263459.1 27 - 28 4566 67
XM_008265239.2 XP_008263461.1 27 - 28 4566 67
XM_008265241.2 XP_008263463.1 27 - 28 4563 67
XM_008265242.2 XP_008263464.1 27 - 28 4550 67
XM_008265243.2 XP_008263465.1 27 - 28 4562 67
XM_008265244.2 XP_008263466.1 27 - 28 4559 67
XM_008265246.2 XP_008263468.1 27 - 28 4546 67
XM_008265247.2 XP_008263469.1 27 - 28 4570 67
XM_008265248.2 XP_008263470.1 26 - 27 4353 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

nucleotidyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0