Assay Details
Gene Symbol: GOLGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719592.3 | XP_002719638.1 | 4 - 5 | 1263 | 57 | |
XM_008271909.2 | XP_008270131.1 | 4 - 5 | 1265 | 57 | ||
XM_017349463.1 | XP_017204952.1 | 4 - 5 | 1265 | 57 | ||
XM_017349464.1 | XP_017204953.1 | 4 - 5 | 1266 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
