This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06738801_m1
See other ZNF7 GE Assays ›
Gene Symbol:
ZNF7
Gene Name:
zinc finger protein 7
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 163879 - 180159 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06738801_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008251914.2 XP_008250136.1 370 66
XM_008251915.2 XP_008250137.1 557 66
XM_008251916.2 XP_008250138.1 367 66
XM_008251917.2 XP_008250139.1 337 66
XM_008251918.2 XP_008250140.1 524 66
XM_008251919.2 XP_008250141.1 227 66
XM_008251923.2 XP_008250145.1 397 66
XM_017339541.1 XP_017195030.1 213 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017339541.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0