Assay Details
Gene Symbol: KCTD20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714660.3 | XP_002714706.1 | 5 - 6 | 820 | 80 | |
XM_008262838.2 | XP_008261060.1 | 3 - 4 | 316 | 80 | ||
XM_017345072.1 | XP_017200561.1 | 5 - 6 | 722 | 80 | ||
XM_017345073.1 | XP_017200562.1 | 5 - 6 | 722 | 80 | ||
XM_017345074.1 | XP_017200563.1 | 4 - 5 | 606 | 80 | ||
XM_017345075.1 | XP_017200564.1 | 4 - 5 | 448 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
