This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06731677_m1
See other LOC100345657 GE Assays ›
Gene Symbol:
LOC100345657
Gene Name:
probable ATP-dependent RNA helicase DDX60
Gene Aliases:
DDX60
Chromosome Location:
Chr.2: 46416761 - 46516561 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06731677_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100345657
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008249573.2 XP_008247795.1 30 - 31 4395 94
XM_008249579.2 XP_008247801.1 30 - 31 4326 94
XM_008249585.2 XP_008247807.1 30 - 31 4389 94
XM_008249590.2 XP_008247812.1 30 - 31 4389 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm