This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06725509_g1
See other ECM1 GE Assays ›
Gene Symbol:
ECM1
Gene Name:
extracellular matrix protein 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 41564250 - 41570890 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06725509_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008264481.2 XP_008262703.1 9 - 10 1880 69
XM_008264482.2 XP_008262704.1 9 - 10 1877 69
XM_008264483.2 XP_008262705.1 10 - 11 1778 69
XM_008264485.2 XP_008262707.1 8 - 9 1505 69
XM_008264486.2 XP_008262708.1 9 - 10 1403 69
XM_017345908.1 XP_017201397.1 9 - 10 1697 69
XM_017345909.1 XP_017201398.1 9 - 10 1694 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0