Assay Details
Gene Symbol: AMMECR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712235.3 | XP_002712281.1 | 3 - 4 | 708 | 61 | |
XM_008258440.2 | XP_008256662.1 | 2 - 3 | 730 | 61 | ||
XM_008258441.2 | XP_008256663.1 | 2 - 3 | 738 | 61 | ||
XM_008258443.2 | XP_008256665.1 | 2 - 3 | 743 | 61 | ||
XM_008258445.2 | XP_008256667.1 | 2 - 3 | 597 | 61 | ||
XM_017342779.1 | XP_017198268.1 | 2 - 3 | 740 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
