This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06704720_m1
See other LOC100344331 GE Assays ›
Gene Symbol:
LOC100344331
Gene Name:
calcium-binding mitochondrial carrier protein SCaMC-1
Gene Aliases:
-
Chromosome Location:
Chr.13: 55710792 - 55758208 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06704720_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100344331
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008265588.1 XP_008263810.1 1 - 2 253 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0