This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06701592_m1
See other GPR89B GE Assays ›
Gene Symbol:
GPR89B
Gene Name:
G protein-coupled receptor 89B
Gene Aliases:
-
Chromosome Location:
Chr.13: 43070590 - 43138586 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06701592_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR89B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002715650.3 XP_002715696.1 6 - 7 667 69
XM_008264554.2 XP_008262776.1 6 - 7 681 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0