This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06696588_m1
See other LOC100349807 GE Assays ›
Gene Symbol:
LOC100349807
Gene Name:
glycerophosphodiester phosphodiesterase domain-containing protein 4
Gene Aliases:
GDPD4
Chromosome Location:
Chr.1: 139416332 - 139538362 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06696588_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100349807
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008263611.2 XP_008261833.1 1 - 2 217 61
XM_017344511.1 XP_017200000.1 1 - 2 217 61
XM_017344512.1 XP_017200001.1 1 - 2 219 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008263611.2, XM_017344511.1, XM_017344512.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

phosphodiesterase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0