This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06686418_g1
See other HAPLN1 GE Assays ›
Gene Symbol:
HAPLN1
Gene Name:
hyaluronan and proteoglycan link protein 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 3879343 - 3959756 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06686418_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAPLN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002713896.3 XP_002713942.1 3 - 4 830 85
XM_008261911.2 XP_008260133.1 4 - 5 810 85
XM_017344570.1 XP_017200059.1 3 - 4 938 85
XM_017344571.1 XP_017200060.1 4 - 5 826 85
XM_017344572.1 XP_017200061.1 4 - 5 757 85
XM_017344573.1 XP_017200062.1 2 - 3 757 85
XM_017344574.1 XP_017200063.1 3 - 4 937 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0