This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06682719_m1
See other S100PBP GE Assays ›
Gene Symbol:
S100PBP
Gene Name:
S100P binding protein
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 5416343 - 5460186 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06682719_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100PBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002720706.3 XP_002720752.1 1316 62
XM_008273740.2 XP_008271962.1 1315 62
XM_008273742.2 XP_008271964.1 1306 62
XM_008273743.2 XP_008271965.1 1260 62
XM_008273745.2 XP_008271967.1 1196 62
XM_017350215.1 XP_017205704.1 1374 62
XM_017350216.1 XP_017205705.1 1259 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0