Assay Details
Gene Symbol: VARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262599.2 | XP_008260821.1 | 11 | 1869 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
