This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06673518_g1
See other RPL26L1 GE Assays ›
Gene Symbol:
RPL26L1
Gene Name:
ribosomal protein L26 like 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 54013959 - 54023967 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06673518_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL26L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002710380.3 XP_002710426.3 1 - 2 129 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_002710380.3 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0