Assay Details
Gene Symbol: CTNNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709686.2 | XP_002709732.1 | 15 - 16 | 2302 | 100 | |
XM_002709687.2 | XP_002709733.1 | 15 - 16 | 2302 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
