This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc03397595_m1
See other KCNMB1 GE Assays ›
Gene Symbol:
KCNMB1
Gene Name:
potassium calcium-activated channel subfamily M regulatory beta subunit 1
Gene Aliases:
BKBETA
Chromosome Location:
Chr.3: 51756479 - 51764200 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc03397595_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNMB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082225.1 NP_001075694.1 3 - 4 756 83
GenBank mRNA AB001934.1 - 3 - 4 352 83
AB009313.1 - 3 - 4 395 83
AF107300.1 - 3 - 4 381 83
AY829265.1 - 3 - 4 373 83
DQ821756.2 - 3 - 4 756 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0