This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc03396131_m1
See other FKBP3 GE Assays ›
Gene Symbol:
FKBP3 LOC100348481
Gene Name:
FKBP prolyl isomerase 3
peptidyl-prolyl cis-trans isomerase FKBP3-like
Gene Aliases:
FKBP25
-
Chromosome Location:
Chr.17: 64490815 - 64503930 on Build OryCun2.0
Chromosome Location:
Chr.8: 75145209 - 75146014 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
-
Assay ID Oc03396131_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FKBP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002718166.3 XP_002718212.1 6 - 7 662 133
GenBank mRNA D82876.1 - 6 - 7 620 133
Gene Symbol: LOC100348481
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017343517.1 XP_017199006.1 1 - 2 648 133

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0