This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04894256_g1
See other TRIR GE Assays ›
Gene Symbol:
TRIR
Gene Name:
telomerase RNA component interacting RNase
Gene Aliases:
C5H19orf43
Chromosome Location:
Chr.5: 11165934 - 11168997 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04894256_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRIR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027969568.1 XP_027825369.1 1 - 2 397 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

DNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0