Assay Details
Gene Symbol: DPY19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095308.2 | XP_014950794.2 | 1 - 2 | 370 | 65 | |
XM_027968631.1 | XP_027824432.1 | 1 - 2 | 370 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
