Assay Details
Gene Symbol: PPP1R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095296.2 | XP_014950782.2 | 3 - 4 | 991 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
