This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04874733_m1
See other ADGRG3 GE Assays ›
Gene Symbol:
ADGRG3
Gene Name:
adhesion G protein-coupled receptor G3
Gene Aliases:
GPR97
Chromosome Location:
Chr.14: 27252830 - 27277964 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04874733_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRG3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027977876.1 XP_027833677.1 4 - 5 548 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Ovis aries

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0