This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04865483_g1
See other HTD2 GE Assays ›
Gene Symbol:
HTD2 RPP14
Gene Name:
hydroxyacyl-thioester dehydratase type 2
ribonuclease P/MRP subunit p14
Gene Aliases:
-
-
Chromosome Location:
Chr.19: 45008675 - 45020641 on Build Oar_rambouillet_v1.0
Chromosome Location:
Chr.19: 45011810 - 45020893 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
-
Assay ID Oa04865483_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HTD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027958244.1 XP_027814045.1 5 - 6 677 65
Gene Symbol: RPP14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027958243.1 XP_027814044.1 4 - 5 438 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_027958244.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Ovis aries

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0