This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04863994_m1
See other DNAJB6 GE Assays ›
Gene Symbol:
DNAJB6
Gene Name:
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases:
-
Chromosome Location:
Chr.4: 128745655 - 128787151 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04863994_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNAJB6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012177402.3 XP_012032792.2 4 - 5 457 77
XM_027969070.1 XP_027824871.1 4 - 5 373 77
XM_027969071.1 XP_027824872.1 4 - 5 1684 77
XM_027969072.1 XP_027824873.1 4 - 5 379 77
XM_027969073.1 XP_027824874.1 4 - 5 373 77
XM_027969074.1 XP_027824875.1 4 - 5 373 77
XM_027969076.1 XP_027824877.1 4 - 5 410 77
XM_027969077.1 XP_027824878.1 4 - 5 339 77
XM_027969078.1 XP_027824879.1 4 - 5 394 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0