Assay Details
Gene Symbol: VARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958818.1 | XP_027814619.1 | 23 - 24 | 2929 | 78 | |
XM_027958819.1 | XP_027814620.1 | 23 - 24 | 2938 | 78 | ||
XM_027958821.1 | XP_027814622.1 | 20 - 21 | 2107 | 78 | ||
XM_027958822.1 | XP_027814623.1 | 23 - 24 | 2531 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
