This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04857370_m1
See other MIA2 GE Assays ›
Gene Symbol:
MIA2
Gene Name:
MIA SH3 domain ER export factor 2
Gene Aliases:
CTAGE5
Chromosome Location:
Chr.18: 48380514 - 48477211 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04857370_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004017912.4 XP_004017961.2 17 - 18 1497 127
XM_012098847.3 XP_011954237.2 23 - 24 3320 127
XM_012098849.3 XP_011954239.2 22 - 23 3293 127
XM_012098850.3 XP_011954240.2 23 - 24 3320 127
XM_012098851.3 XP_011954241.2 22 - 23 3293 127
XM_012098852.3 XP_011954242.2 18 - 19 1446 127
XM_012098853.3 XP_011954243.2 18 - 19 2514 127
XM_012098854.3 XP_011954244.2 18 - 19 1524 127
XM_012098855.3 XP_011954245.2 18 - 19 2394 127
XM_012098857.3 XP_011954247.2 18 - 19 2385 127
XM_012098858.3 XP_011954248.2 17 - 18 1497 127
XM_027957293.1 XP_027813094.1 18 - 19 1524 127
XM_027957294.1 XP_027813095.1 18 - 19 2513 127
XM_027957295.1 XP_027813096.1 18 - 19 2385 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Ovis aries

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0