This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04853578_g1
See other CEP43 GE Assays ›
Gene Symbol:
CEP43
Gene Name:
centrosomal protein 43
Gene Aliases:
FGFR1OP
Chromosome Location:
Chr.8: 96522170 - 96550057 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04853578_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEP43
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015097272.2 XP_014952758.2 12 - 13 1170 111
XM_027972658.1 XP_027828459.1 11 - 12 1169 111
XM_027972659.1 XP_027828460.1 9 - 10 948 111

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0