This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04852195_g1
See other LOC101114075 GE Assays ›
Gene Symbol:
LOC101114075
Gene Name:
sulfotransferase 1A1
Gene Aliases:
SULT1A1
Chromosome Location:
Chr.24: 26658418 - 26662679 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04852195_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101114075
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004020871.4 XP_004020920.1 5 - 6 1104 97
XM_012104340.3 XP_011959730.1 5 - 6 1140 97
XM_012104341.3 XP_011959731.1 4 - 5 893 97
XM_012104342.3 XP_011959732.1 5 - 6 1100 97
XM_012104344.3 XP_011959734.1 4 - 5 929 97
XM_012104345.3 XP_011959735.1 4 - 5 889 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Ovis aries

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0