Assay Details
Gene Symbol: GCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019134.4 | XP_004019183.1 | 1 - 2 | 93 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: GCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019134.4 | XP_004019183.1 | 1 - 2 | 93 | 64 |
Set Membership: |
![]() ![]() ![]() |