Assay Details
Gene Symbol: WBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013122.4 | XP_004013171.2 | 2 - 3 | 336 | 62 | |
XM_027974093.1 | XP_027829894.1 | 2 - 3 | 294 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
