This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04825264_g1
See other LOC105607811 GE Assays ›
Gene Symbol:
LOC105607811
Gene Name:
protein phosphatase inhibitor 2-like
Gene Aliases:
-
Chromosome Location:
Chr.11: 36314818 - 36318814 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04825264_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105607811
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027974985.1 XP_027830786.1 3 - 4 738 102
XM_027974986.1 XP_027830787.1 3 - 4 738 102
XM_027974987.1 XP_027830788.1 2 - 3 630 102
XM_027974988.1 XP_027830789.1 3 - 4 737 102
XM_027974989.1 XP_027830790.1 3 - 4 735 102
XM_027974990.1 XP_027830791.1 4 - 5 436 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Ovis aries

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase inhibitor phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0