This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04824033_g1
See other PRKRA GE Assays ›
Gene Symbol:
PRKRA
Gene Name:
protein activator of interferon induced protein kinase EIF2AK2
Gene Aliases:
-
Chromosome Location:
Chr.2: 142291259 - 142306931 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04824033_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRKRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004004546.4 XP_004004595.1 4 - 5 527 70
XM_012132652.3 XP_011988042.2 4 - 5 527 70
XM_027964797.1 XP_027820598.1 2 - 3 97 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0