Assay Details
Gene Symbol: ENC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980005.1 | XP_027835806.1 | 2 - 3 | 2190 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: ENC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980005.1 | XP_027835806.1 | 2 - 3 | 2190 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() |