Assay Details
| Gene Symbol: SDR42E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977666.1 | XP_027833467.1 | 2 - 3 | 169 | 70 | |
| XM_027977667.1 | XP_027833468.1 | 2 - 3 | 1890 | 70 | ||
| XM_027977668.1 | XP_027833469.1 | 3 - 4 | 1266 | 70 | ||
More Information
Set Membership: |
|

SDR42E1
Genomic Map