This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04785050_m1
See other AKTIP GE Assays ›
Gene Symbol:
AKTIP
Gene Name:
AKT interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.14: 23329508 - 23339729 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04785050_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKTIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004014986.4 XP_004015035.1 3 - 4 443 61
XM_012189780.3 XP_012045170.1 3 - 4 470 61
XM_012189781.3 XP_012045171.1 4 - 5 572 61
XM_012189783.3 XP_012045173.1 3 - 4 453 61
XM_012189784.3 XP_012045174.1 3 - 4 597 61
XM_012189785.3 XP_012045175.1 3 - 4 484 61
XM_027977844.1 XP_027833645.1 4 - 5 557 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0