Assay Details
Gene Symbol: MINDY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002469.3 | XP_004002518.2 | 5 - 6 | 1661 | 56 | |
XM_012180804.2 | XP_012036194.2 | 6 - 7 | 1288 | 56 | ||
XM_012180810.2 | XP_012036200.2 | 5 - 6 | 1528 | 56 | ||
XM_012180814.2 | XP_012036204.2 | 6 - 7 | 1267 | 56 | ||
XM_012180827.3 | XP_012036217.2 | 6 - 7 | 1323 | 56 | ||
XM_027974581.1 | XP_027830382.1 | 5 - 6 | 1507 | 56 | ||
XM_027974587.1 | XP_027830388.1 | 5 - 6 | 1507 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
