Assay Details
Gene Symbol: LOC114112016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963702.1 | XP_027819503.1 | 2 - 3 | 278 | 91 |
Gene Symbol: SWT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187729.3 | XP_012043119.1 | 13 - 14 | 2173 | 91 | |
XM_012187733.3 | XP_012043123.1 | 13 - 14 | 2173 | 91 |
