Assay Details
Gene Symbol: RBM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960455.1 | XP_027816256.1 | 13 - 14 | 3760 | 53 | |
XM_027960456.1 | XP_027816257.1 | 13 - 14 | 3796 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
