This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04730960_g1
See other SLC16A3 GE Assays ›
Gene Symbol:
SLC16A3
Gene Name:
solute carrier family 16 member 3
Gene Aliases:
-
Chromosome Location:
Chr.11: 12460782 - 12472317 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04730960_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC16A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004013053.4 XP_004013102.1 3 - 4 488 65
XM_012186383.3 XP_012041773.1 4 - 5 612 65
XM_012186384.3 XP_012041774.1 4 - 5 1619 65
XM_012186387.3 XP_012041777.1 3 - 4 1213 65
XM_012186389.3 XP_012041779.1 3 - 4 3230 65
XM_027974308.1 XP_027830109.1 4 - 5 1473 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0