This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04730812_m1
See other AKT2 GE Assays ›
Gene Symbol:
AKT2
Gene Name:
AKT serine/threonine kinase 2
Gene Aliases:
-
Chromosome Location:
Chr.14: 53022208 - 53071015 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04730812_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027978322.1 XP_027834123.1 9 - 10 1077 68
XM_027978323.1 XP_027834124.1 11 - 12 3399 68
XM_027978324.1 XP_027834125.1 9 - 10 1073 68
XM_027978325.1 XP_027834126.1 11 - 12 1350 68
XM_027978326.1 XP_027834127.1 10 - 11 1321 68
XM_027978327.1 XP_027834128.1 9 - 10 1151 68
XM_027978328.1 XP_027834129.1 10 - 11 1213 68
XM_027978329.1 XP_027834130.1 9 - 10 1007 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0