Assay Details
Gene Symbol: RSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965498.1 | XP_027821299.1 | 2 - 3 | 728 | 65 | |
XM_027965499.1 | XP_027821300.1 | 2 - 3 | 724 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
