Assay Details
Gene Symbol: VSIG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956534.1 | XP_027812335.1 | 2 - 3 | 633 | 73 | |
XM_027956535.1 | XP_027812336.1 | 2 - 3 | 638 | 73 | ||
XM_027956536.1 | XP_027812337.1 | 2 - 3 | 645 | 73 | ||
XM_027956537.1 | XP_027812338.1 | 2 - 3 | 648 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
