This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04711933_m1
See other ATP6AP1L GE Assays ›
Gene Symbol:
ATP6AP1L
Gene Name:
ATPase H+ transporting accessory protein 1 like
Gene Aliases:
-
Chromosome Location:
Chr.5: 86105362 - 86129285 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04711933_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP6AP1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004009062.3 XP_004009111.3 4 - 5 547 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0