This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04678060_m1
See other LOC101110111 GE Assays ›
Gene Symbol:
LOC101110111
Gene Name:
selenocysteine lyase
Gene Aliases:
SCLY
Chromosome Location:
Chr.1: 3100860 - 3130948 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04678060_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101110111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027967129.1 XP_027822930.1 4 - 5 561 80
XM_027967134.1 XP_027822935.1 4 - 5 521 80
XM_027967140.1 XP_027822941.1 4 - 5 464 80
XM_027967147.1 XP_027822948.1 4 - 5 506 80
XM_027967151.1 XP_027822952.1 4 - 5 495 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Ovis aries

Panther Classification:

Molecular Function -

lyase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0