This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04671333_m1
See other LOC101123244 GE Assays ›
Gene Symbol:
LOC101123244
Gene Name:
serpin B9
Gene Aliases:
SERPINB9
Chromosome Location:
Chr.20: 54278887 - 54286764 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04671333_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101123244
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012101374.3 XP_011956764.2 5 - 6 678 84
XM_027958714.1 XP_027814515.1 5 - 6 672 84
XM_027958715.1 XP_027814516.1 5 - 6 670 84
XM_027958716.1 XP_027814517.1 5 - 6 676 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Ovis aries

Panther Classification:

Molecular Function -

protease inhibitor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0