Assay Details
Gene Symbol: SCGB1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178105.1 | NP_001171576.1 | 2 - 3 | 237 | 65 | |
GenBank mRNA | FJ959385.1 | - | 2 - 3 | 237 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
